Would you like to log in again?
HiVeg™ Columbia Broth Base, HiMedia
Supplier: HiMedia Laboratories
HiVeg™
MV145-500G MV145-500G
61000-682EA 226.56 USD InStock
CA61000-682 61000-682
HiVeg™ Columbia Broth Base, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Columbia broth base are recommended for the cultivation of fastidious organisms from clinical specimens.
This medium is prepared by completely replacing animal based peptones with vegetable peptones. In this medium the growth appears earlier in a substantial number of clinical blood cultures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty