Would you like to log in again?
HiVeg™ Tributyrin Agar Base, HiMedia Laboratories
Supplier: HiMedia
MV157-500G MV157-500G MV157-2.5KG MV157-5KG MV157-2.5KG MV157-5KG
89228-114EA 1213.74 USD InStock
CA89228-114 CA89228-112 89228-114 89228-112 CA61000-694 61000-694
HiVeg™ Tributyrin Agar Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Used for the detection of lipolytic microorganisms.
  • Light yellow colored
  • Opalescent gel forms, with oil droplets in petri plates
  • Reaction of 2.3% w/v aqueous solution containing 1% v/v tributyrin at 25 °C
Caution: Store the dry medium below 30 °C and the prepared medium at 2 to 8 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty