Would you like to log in again?
Tetrathionate HiVegTM Broth Base, Hajna, HiMedia Laboratories
Supplier: HiMedia Laboratories
MV327-500G MV327-500G
61000-892EA 114.9 USD InStock
CA61000-892 61000-892
Tetrathionate HiVegTM Broth Base, Hajna, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
TT HiVeg Broth base is the modification of TT Broth formulated according to Hajna and Damon who modified the medium described by Kauffmann and Knox HiVeg special peptone and yeast extract provide nitrogenous compounds, vitamin B complex and other essential nutrients.
  • Uses HiVeg special peptone and synthetic detergent No. III
  • Inhibit gram-positive bacteria
  • Formed in the medium by the addition of a solution containing iodine and potassium iodide
Tetrathionate HiVeg Broth Base is developed by using HiVeg special peptone and synthetic detergent No. III which is free of BSE/TSE risks associated with animal based ingredients. TT HiVeg Broth base is the modification of TT Broth formulated according to Hajna and Damon who modified the medium described by Kauffmann and Knox HiVeg special peptone and yeast extract provide nitrogenous compounds, vitamin B complex and other essential nutrients. Synthetic detergent No. III and brilliant green inhibit gram-positive bacteria. Dextrose and mannitol are the fermentable carbohydrates which help in differentiation of the enteric pathogens especially
Salmonella group. The combination of sodium thiosulphate and tetrathionate helps in suppressing coliform organisms making this medium more selective. Tetrathionate is formed in the medium by the addition of a solution containing iodine and potassium iodide. Sodium chloride maintains the osmotic balance of the medium whereas calcium carbonate is a neutralizer that absorbs toxic metabolites. After enrichment, the growth may be streaked for further confirmation on MacConkey HiVeg Agar.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty