Would you like to log in again?
HiVeg™ Bennet’s Agar, HiMedia
Supplier: HiMedia Laboratories
MV694-2.5KG MV694-2.5KG MV694-500G MV694-5KG MV694-5KG MV694-500G
89229-052EA 1446.73 USD InStock
CA89229-052 89229-052 CA89229-050 CA61001-244 89229-050 61001-244
HiVeg™ Bennet’s Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Bennet’s HiVeg agar is used for cultivation and enhancement of sporulation of Nocardia and Streptomyces.
  • Light yellow coloured, may have slightly greenish tinge, homogeneous, free flowing powder
  • Storage below 30 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty