Would you like to log in again?
Wilkins Chalgren Anaerobic HiVegTM Agar/Broth Base, HiMedia Laboratories
Supplier: HiMedia Laboratories
MV832-500G MV863-500G MV832-500G MV863-5KG MV863-5KG MV863-500G MV832-5KG MV863-2.5KG MV832-2.5KG MV863-2.5KG MV832-2.5KG MV832-5KG
89229-262EA 780.54 USD InStock
CA89229-264 89229-262 CA89229-262 89229-264 CA61001-342 89229-192 CA61001-376 CA89229-192 CA89229-190 61001-342 89229-190 61001-376
Wilkins Chalgren Anaerobic HiVegTM Agar/Broth Base, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Wilkins Chalgren Anaerobic HiVeg agar/broth base is used for the isolation and cultivation of anaerobic bacteria and also for susceptibility testing of anaerobic bacteria by the agar dilution method.
  • Appearance of powder: Yellow colored, slightly greenish tinge, homogeneous, free flowing powder
  • Colour and clarity: Medium amber colored, clear to slightly opalescent gel forms in petri plates, clear solution in tubes
Wilkins Chalgren HiVeg Media are prepared by replacing animal based peptones of bovine origin by vegetable peptones. The media have yeast extract to provide vitamins and other growth factors like purines and pyrimidines that are essential for the growth of P. melaninogenica. Arginine serves as an amino acid source especially for Eubacterium lentum and pyruvate is an energy source for Asaccharolytic cocci such as Veillonella. Pyruvate with arginine can provide proper environment for anaerobic growth. Ferric pyrophosphate is necessary for the growth of Bacteroides species and menadione for P. melaninogenica.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty