Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Meat Extract Powder, HiMedia

Supplier: HiMedia Laboratories
RM003-500G RM003-500G
61001-514EA 128.54 USD InStock
61001-514 CA61001-514
Meat Extract Powder, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Meat extract powder is prepared from fresh lean meat specially selected to yield maximum growth of fastidious microorganisms

  • Animal containing
  • Yellowish brown coloured powder having an agreeable meat like odour and taste
  • Readily dissolves in distilled water and remains clear even after autoclaving
  • Insoluble in alcohol
  • Animal containing
  • Reaction of 2% w/v aqueous solution at 25 °C
  • pH range - 5.90 - 6.90

The high quality pure lean meat used for this, ensures consistent colony characteristics, when used in differential media. The media has to be stored below 30°C.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty