Would you like to log in again?
Bile, Dried, Purified, HiMedia
Supplier: HiMedia Laboratories
RM010-10KG RM010-2.5KG RM010-500G RM010-2.5KG RM010-25KG RM010-5KG RM010-10KG RM010-5KG RM010-25KG RM010-500G
89229-584EA 7507.12 USD InStock
CA89229-584 89229-584 89229-586 CA61001-534 CA89229-590 89229-590 61001-534 CA89229-588 89229-588 CA89229-586
Bile, Dried, Purified, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Bile is obtained from purified fresh bile dessicated under controlled conditions of temperature that helps to ensure a uniform and reproducible product
  • Greenish yellow, homogeneous, free flowing powder having bile like odour
  • Final pH: 6.50 - 7.50
A selectively inhibitory agent in microbiological culture media.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty