Would you like to log in again?
Casein Enzyme Hydrolysate, Type - II, HiMedia
Supplier: HiMedia Laboratories
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
RM028-500G RM028-25KG RM028-500G RM028-2.5KG RM028-5KG RM028-10KG
89229-718EA 1661.27 USD InStock
89229-718 89229-714 CA61001-574 89229-716 89229-712 61001-574
Casein Enzyme Hydrolysate, Type - II, HiMedia
Media for Microbiology Additives and Supplements for Microbiology
This is equivalent to Casitone and is used in antibiotic assay media.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty