Would you like to log in again?
ARMOR 12 INCH
Supplier: G&W INSTRUMENTS
Thermometer Armor^ 30.5cm open-faced armor that is brass-nickel plated. For use with a 30.5cm thermometer.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ARMOR 12 INCH 61017-561 7005 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty