Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Digital Outlet Controller

Supplier: VWR
VWR®
93678-81
61161-343EA 101.81 USD InStock
61161-343
VWR® Digital Outlet Controller
Timers Time Switches
Versatile and economical controller programs equipment on and off at any minute of any day.

  • Traceable®
  • High-impact ABS plastic case
  • Motor load capacity: 1 hp

A fail-safe memory permits programming 14 different switchings per day.

The 1.3 cm (¹/₂") LCD shows the time-of-day, the day of the week, and the event number of the program taking place. Manual override button permits equipment to be switched on/off independent of event settings. Program and clock times are maintained by a backup battery in the event of an electrical outage.

They may be allocated over four separate weekly programs for up to 196 switchings per week. Controller will turn equipment on and off in intervals as short as 1 minute or as long as 168 hours. The a.m./p.m. day programs allow repeating the same cycle every day of the week, weekdays only, weekends only, or any selected days.

Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Ordering information: Includes two 1.5 V batteries and operating instructions.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty