Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® Thermometer/Humidity Wall Clock

Supplier: Avantor
VWR®
90027-87
61161-348EA 77.19 USD InStock
61161-348
VWR® Thermometer/Humidity Wall Clock
Timers Wall Clocks
Lab wall clock displays time, temperature, and humidity.

  • Traceable®
  • Bright red coloring
  • Highly visible
  • Accurate time display

Bright red sweeping second hand provides superior visibility. Timing accuracy is 0.01%.

The 31.8 cm (12¹/₂") dial displays quartz-crystal timing.

Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Ordering information: Supplied with AA alkaline battery.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty