Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Dial Thermometers
Supplier: VWR
VWR®
61018-52
61220-418EA 71.15 USD InStock
61220-418
VWR® Dial Thermometers
Thermometers Digital Thermometers
Thermometer is used to measure temperature of gases, food, and other semisolids.
  • Available in °C and °F
  • Less weighed thermometer
Longer stem is excellent for measuring sample temperatures while stirring in deep vessels or water baths.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty