Would you like to log in again?
SC3 Super Compact Temperature Recorders, Dickson®
Supplier: Dickson
SC386 C314 C313 C310 C316 C311 C312 SC397 SC377 C317 SC387 SC367 C315 C318 C309
14905-004EA 598.86 USD InStock
14905-004 14905-008 61280-490 14905-006 61280-040 61280-028 61280-547 61280-032 61280-020 61280-034 61280-540 61280-044 61280-024 14905-012 14905-010
SC3 Super Compact Temperature Recorders, Dickson®
Chart Recorders Recorders
Reusable and compact recorders are built with a rugged case to withstand the rigors of transportation and storage applications
Economical recorders feature easy user calibration and switchable control of temperature, °C/°F, and time, 24 hours/7 days. Designed with specialized slots for unit security. Optional tamper seals can be used to ensure the integrity of the chart. Units can be wall-mounted, suspended, or used free-standing, and work with charts that have a diameter of 75mm (3").
Ordering information: Supplied with one AA battery, pen, and instruction manual.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty