Would you like to log in again?
KT8 Thermocouple Remote Sensing Chart Recorders, Dickson®
Supplier: Dickson
C674 C444 C440 C671 C435 C408 C307 C412 C675 C303 C407 C405 C441 C676 C410 C442 C439 C302 C670 C424 C463 C420 C655 C452 C177 C669 C319 C101 C668 C304 KT8P2 C443 C308 C432 KT8P3 C428 C498 C672 C673 C320 C409 C306 C431 C414 C422 C459 C406 C301 C305 C457 C495 C411
61280-030EA 106.4 USD InStock
61280-030 61280-470 61280-509 61280-512 61280-036 61280-476 61280-478 61280-510 61280-480 61280-403 61280-644 61280-405 61280-646 61280-407 61280-648 61280-409 61280-482 61280-042 61280-485 61280-487 61280-046 61280-640 61280-563 61280-400 61280-488 61280-656 61280-018 61280-413 61280-658 61280-415 61280-538 61280-130 61280-650 61280-451 61280-652 61280-453 61280-456 61280-654 61280-455 61280-411 61280-468 61280-504 61279-924 61279-922 61280-507 61280-660 61280-022 61280-662 61280-544 61280-026 61280-224 61280-620
KT8 Thermocouple Remote Sensing Chart Recorders, Dickson®
Chart Recorders Recorders
These chart recorders do remote monitoring of temperatures in warehouses, food processing facilities, archives, laboratories, and blood storage, dialysis and medical centers.
  • Wall mountable
  • Flip-up pen arm allows for easy chart and pen changes
  • K-thermocouple probe for flexibility, user-selectable temperature ranges and recording times
  • 9V battery backup
Recorders have a rugged, compact enclosure with an ABS case, polycarbonate door, and aluminum dial. The large 20.3 cm (8") charts (sold separately) offer greater resolution. These data acquisition instruments are ideal for remote monitoring of temperatures in refrigerators, freezers, coolers, ovens, smokers, autoclaves, baths, cleanrooms, chambers, storage rooms, air ducts, vents, and critical equipment storage. Units 61279-922, -924, and -926 feature 2.5 cm (1") 3.5 digit LCD, viewable from 7.6 m (25') and Units 61279-924 and -926 also feature a high/low audio/visual alarm.
Certifications: CE marked.
Ordering information: Units supplied with 120 V adapter, pens, and 9 V battery. The 61279-922 and -924 include one 1.2 m (4') bead wire K-thermocouple probe. The 61279-926 includes a 15.2 cm (6") high temperature basic K-thermocouple probe. Charts must be ordered separately.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty