Would you like to log in again?
Corning® Tris Base Buffer, Powder, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Corning®
61-233-RM 61-233-RT 61-233-RT 61-233-RR 61-233-RR 61-233-RM
62111-160EA 973.17 USD InStock
62111-160 CA62111-160 62111-162 CA62111-162 62111-158 CA62111-158
Corning® Tris Base Buffer, Powder, Corning
Buffers Electrophoresis Buffers Running Buffers
Tris is commonly used in buffer solutions in biochemical and molecular biological research
This primary amine has an effective pH range of 7.0 - 9.2 and is reported to inhibit a number of different enzymes, limiting its usefulness in protein studies to some extent. This tris base buffer is provided in powder form.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty