Would you like to log in again?
Eppendorf® UVette® Disposable Cuvettes
Supplier: Eppendorf
4099005004 952010077 952010051 952010069 952010123 952010051 952010069 952010131 952010077 952010115 952010107
47735-408EA 230.16 USD InStock
47735-408 89094-396 CA47735-408 CA89094-396 75874-874 CA35920-109 62111-470 62111-472 62111-474 47735-412 62111-412
Eppendorf® UVette® Disposable Cuvettes
Cuvettes Spectrophotometer Cuvettes
These disposable cuvettes allow for measurements in the UV/VIS range with transparency from 230nm to 1600nm
Choose between optical path lengths of 10mm or 2mm with a single 90° rotation. The UVette® is ideally matched for use with the Eppendorf® BioPhotometer® Plus and BioSpectrometer®. For use with other spectrophotometers, the UVette® can be utilized with the aid of an adapter.
Ordering information: The UVette® Routine Pack is supplied in a reclosable box. The UVette® Starter Set includes 80 UVettes and one universal adapter for 8.5mm and 15mm light path heights.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty