Would you like to log in again?
neodisher® Detergents, Miele
Supplier: Miele
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
neodisher®
94069007 94069005 94000004 94069001 94000005 94069002 94069014 94069010 94069003 94069008 94069017 94069016 94000090 94069004 94069011
62111-844EA 382.17 USD InStock
62111-844 62111-834 62111-856 62111-846 62111-836 62111-858 62111-848 62111-838 62111-683 62111-840 62111-686 62111-830 62111-852 62111-842 62111-832
neodisher® Detergents, Miele
Cleaning Agents Detergents and Surface Cleansers
These powder and liquid detergents provide consistent, high level cleanliness in Miele glassware washing systems
Detergents FA and MA are for the removal of creams and lotions, cleaning stainless steel, and washing dental instruments. Detergents FT and F are for the removal of inks. Detergent FT, as well as GK, is also used for cell and tissue culture residues. Detergent FLA works well with wastewater, food, environmental, educational, and chemical applications. Detergent A8 is suited for generalized cleaning; detergent LA removes fats, oils, and waxes in food applications; and detergent UW is suitable for trace metal removal from glassware. Neutralizing agent N is composed of phosphoric and organic acids, while agent Z is composed only of organic acids. Both are meant to be dispensed into the rinse water after the use of alkaline-based detergents.
Ideal for the removal of organic, inorganic, and metal residues from laboratory glassware and medical and dental instruments in many industries, including cosmetic, wastewater, environmental, biotech, and food.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty