Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Traceable® Three-Channel Alarm Timer
Supplier: VWR
VWR®
90009-50
62344-600EA 187.39 USD InStock
62344-600
VWR® Traceable® Three-Channel Alarm Timer
Timers Digital Timers
Time three separate lab operations simultaneously with this unit that also serves as a stopwatch and a time-of-day clock.
  • Perfect unit for repetitive tests
  • Triple display shows three different times at once
  • Tests can be started individually or simultaneously
  • Rugged, chemical-resistant ABS plastic case
  • Timing capacity to 99 hours, 59 minutes and 59 seconds with a 1-second resolution
Channels may be started individually or simultaneously
Adjustable alarm volume allows for use in either a noisy or quiet environment. Alarm may be set to silence automatically after one minute or to sound continuously until turned off. The LCD digits are 9.5 mm (³/₈") high and the display returns to the previously programmed time at the touch of a button. Quartz crystal accuracy is 0.001%.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a one-year, AAA alkaline battery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty