Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Traceable® Four-Channel Alarm Timer with Clock
Supplier: VWR
VWR®
90009-61
62344-641EA 83.04 USD InStock
62344-641
VWR® Traceable® Four-Channel Alarm Timer with Clock
Timers Digital Timers
Microcomputer chip permits setting four separate channels for simultaneous countup or countdown procedures.
  • Massive ³/₄" high display allows viewing from across the room
  • Extra-loud, high decibel alarms for 1 minute and may be silenced manually
  • Timing capacity to 99 hours, 59 minutes and 59 seconds with a 1 second resolution
  • Remarkable memory returns display to previously programmed countdown time at the touch of a button
Large 1.9 cm LCD permits viewing from across the lab. Additional features include stopwatch, time-out, and time-of-day display. Use all four channels simultaneously as count-up stopwatches or in any combination of countup/countdown. The touch of a button returns the display to the previously programmed countdown time. Extra-loud, high-decibel alarm sounds for one minute or may be silenced manually. Finger-size keys make it easy to set times and change channels. Magnetic back allows timer to be placed on any metal surface. Unit also has a flip-open easel for standing on the lab bench, a spring fastener for clipping to a notebook/clipboard, and a hole for use with a lanyard.
When zero is reached the timer starts counting up to show time elapsed since alarming.
Ordering information: Supplied with a one-year, 1.5 V silver-oxide battery. Replacement battery is available separately.
Delivery information: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty