Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Time-Switch Controller
Supplier: VWR
VWR®
5050
62344-723EA 147.02 USD InStock
62344-723
VWR® Time-Switch Controller
Timers Time Switches
Program controller turns equipment on and off at any minute of any day in intervals from 1 minute to 168 hours.
  • Traceable®
  • Program up to 196 on/off changes per week
  • Manual override for user control
Six switches on the front panel control the entire operation
Bright 1.3 cm (¹/₂") high LCD shows the exact time of day complete with a.m./p.m. indicator, day of week, and number of program taking place. A red LED indicates the on/off mode of attached equipment. Housed in high-impact ABS plastic case.
An 'always remember' memory permits programming of 14 different switchings per day, up to 196 unique on/off changes per week. Repeat mode allows programming the same cycle each day for 365 days, repeating a weekly program cycle for 52 weeks, jumping weekends, or skipping any selected days. Manual override permits equipment to be switched on or off immediately without disturbing the program. Fail-safe circuitry ensures the program cannot be erased accidentally. Memory chip stores time settings and clock for up to 30 days in event of power outage.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology. UL listed and CSA certified.
Ordering information: Supplied with AA alkaline battery, operating instructions, High Reliability Certificate, and a form for easy record-keeping.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty