Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® QC Timer™ Timer
Supplier: VWR
VWR® QC Timer™
93678-60
62344-902EA 47.63 USD InStock
62344-902
VWR® QC Timer™ Timer
Timers Digital Timers
Ideal quality control countdown and alarm timer combines easy three-button operation with large 1.9 cm (³/₄") digits.
  • Traceable®
  • Ideal ISO 9000 timer
  • Countdown alarm
  • Magnet/clip on back
Can be set up to 19 hours, 59 minutes. Alarm sounds for one minute.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with three-year AAA alkaline battery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty