Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Lab-Top Timer
Supplier: VWR
VWR®
93678-37
62344-910EA 84.4 USD InStock
62344-910
VWR® Lab-Top Timer
Timers Digital Timers
Designed for bench-use, this timer features any-angle viewing, large keys, and three timing modes.
  • Traceable®
  • For bench use
  • Can display countdown, stopwatch, or time of day
Counts down from 24 hours to 1 second in one second increments. Countdown mode memory allows programming an 'instant replay' repeat time at the touch of a key. Also features a stopwatch and a time-of-day clock.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a three-year AA alkaline battery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty