Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Traceable® Three-Line Alarm Timer
Supplier: VWR
VWR®
90009-69
62344-912EA 97.35 USD InStock
62344-912
VWR® Traceable® Three-Line Alarm Timer
Timers Digital Timers
May be used as three countdown timers, three stopwatches or any combination of up/down timing.
  • Traceable®
  • LCD readout
  • Can display countdown alarm, stopwatch, or time-of-day
  • Separate channels can be started together or individually
Count up or down to 99 hours, 59 minutes, 59 seconds
The three separate channels may be started individually or concurrently. When zero is reached an alarm sounds, 'Time’s Up' is displayed, and the timer begins counting up. Returns to the previously programmed time for repetitive tests.
Also features a time-of-day clock and a magnet/stand back.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with an 18-month, 1.5 V silver-oxide battery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty