Would you like to log in again?
VWR® Traceable® Mini-Thermometer
Supplier: VWR
VWR®
90029-05
62344-925EA 91.6 USD InStock
62344-925
VWR® Traceable® Mini-Thermometer
Thermometers Digital Thermometers
Ideal for measuring liquids, semi-solids, or air/gas, this small powerful unit reads from -50 to 150°C or -58 to 302°F.
  • Traceable®
  • Reads °C and °F
  • Compact size
Switch between °C and °F with the touch of a button. Hold button freezes the display to capture current reading. Readings are updated every second for “no-wait” results. Unit runs for one year on 1.5V silver-oxide battery (included).
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty