Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Professional Time Switches, Capewell Aerial Systems LLC
Supplier: Capewell Aerial Systems
78305 78304
62362-000EA 294 USD InStock
62362-000 62363-003
Professional Time Switches, Capewell Aerial Systems LLC
Timers Time Switches
Time switch has built-in three-wire, grounded plug and receptacle.
  • Built-in grounded plug
  • Marked in one-minute intervals
Time switch has built-in three-wire, grounded plug and receptacle to automatically turn off an electrical device at the end of a selected time period.
Certifications: UL listed.
Ordering information: This product has a minimum order quantity of five units.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty