Would you like to log in again?
Five-Channel Timer, Sper Scientific
Supplier: Sper Scientific
810015 810015C
62364-186EA 156.47 USD InStock
62364-186 62364-185
Five-Channel Timer, Sper Scientific
Timers Digital Timers
This digital count down/count up timer has four independent channels, each of which counts up or down for any interval from 1 second to 100 hours, plus clock button.
  • Extra large display indicates channels in use
  • Each one has a distinctive 1 minute alarm
Automatically begins to count up to indicate time elapsed after count down reaches zero. Memory recalls last timer setting or reuse. Also has a clock mode. Has a stand, pocket clip, and magnet.
Certifications: N.I.S.T. traceable certificate of compliance for the certified option.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty