Would you like to log in again?
Large Display Stopwatch, Sper Scientific
Supplier: Sper Scientific
810022 810022C
62370-038EA 168.44 USD InStock
62370-038 62370-037
Large Display Stopwatch, Sper Scientific
Timers Digital Stopwatches
Large display digital stopwatch features click stop controls, which give the definitive feel of a mechanical stopwatch but with hundreds of times the accuracy.
  • Large LCD for good visibility
  • Rugged ABS plastic case
  • Built-in stand
  • Split interval, clock, and calendar functions
Counts to 24 hours in ¹/₁₀₀th second resolution for the first 30 minutes.
Certifications: N.I.S.T. traceable certificate of compliance for 62370-038.
Ordering information: Leatherette case and One AA battery included. Five Year Warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty