Would you like to log in again?
60 Memory Stopwatch, Sper Scientific
Supplier: Sper Scientific
810033C 810033
62370-047EA 124.02 USD InStock
62370-047 62370-048
60 Memory Stopwatch, Sper Scientific
Timers Digital Stopwatches
Water resistant digital stopwatch features two ¹/₁₀₀th second displays.
  • Counts up or down to 10 hours
  • Ideal for multiple and simultaneous processes
  • Water resistant
In stopwatch mode current lap time shown in upper display, cumulative total in lower one. Memory stores last 60 lap times so there is no need to look away from a test to make notes. It shows fastest, slowest, and average lap times, and has an alarm clock function. In timer mode upper display shows original setting while lower one counts down. Countdown cycles repeat upon completion. 30 second shuttle timer for use during timeouts.
Lap counter shows number of laps (up to 99) in stopwatch mode and number of cycles repeated in timer mode. Pacesetter function can be set to beep continually from 10 beeps per minute to 320 beeps per minute. Shock resistant, superb hand grip.
Certifications: N.I.S.T. traceable certificate of compliance for 62370-048.
Ordering information: Five year warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty