Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

GraLab® Timers, Dimco-Gray

Supplier: Dimco-Gray
GraLab®
7-171-250R 7-173-160R 7-170-160R
62373-109CS 1602 USD InStock
62373-109 62371-040 62371-046
GraLab® Timers, Dimco-Gray
Timers Mechanical Timers
Designed for sustained accuracy required in industrial and laboratory timing applications.

  • A buzzer alarm with variable volume control signals the end of the selected time period
  • Gray polycarbonate/PET case has rubber base pads, wall-mounting keyholes, and a recessed grip for portability

Three ranges available: 60 minutes of timing by minutes and seconds in one-second increments; 15 hours of timing by hours, minutes, and ¹/₄ minutes in 15-second increments; or 60 hours of timing by hours and minutes in one-minute increments.

Large, 16.5 cm (6¹/₂") dial features an outer circle of numerals for interval time and an inner circle of smaller numerals for elapsed time. A three-wire outlet receptacle controls small appliances (<600 W) automatically by a separate switch that can be set to make or break the circuit at zero. Models 62371-045, 62371-046, 62373-109, and 62371-040 add a second three-wire outlet receptacle, independent of the selector switch, which always breaks the circuit at zero.

Max. lamp load: 600 W, power rating of AC outlets (120 V)
Max. motor load: 249 W (¹/₃ hp), power rating of AC outlets (120 V)
Max. resistive load: 1200 W, power rating of AC outlets (120 V)

Certifications: UL listed.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty