Would you like to log in again?
VWR® 60 Memory, Multi-Function Stopwatch with Countdown
Supplier: Avantor
VWR®
90027-78
62379-200EA 102.51 USD InStock
62379-200
VWR® 60 Memory, Multi-Function Stopwatch with Countdown
Timers Digital Stopwatches
Provides repeat functions of countdown alarm timing from 10 hours to 1 second.
  • Traceable®
  • 60 memory settings
  • Programmable
When zero is reached, the timer alarms and begins to count down again from the programmed time.
Three-line display simultaneously shows running time, cumulative split, and interval split. Sixty memories recall all splits, display sample numbers 1 through 60, and show longest, shortest, and average interval splits. Stopwatch times to 9 hours, 59 minutes, 59 seconds, 99 hundredths.
Repeat cycle continues automatically until stopped. Display continually shows running time remaining to zero and cycles counted from 1 to 60.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with lanyard, instructions, and two-year, 3 V lithium battery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty