Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® Extra-Large LCD Digital Stopwatches

Supplier: Avantor
VWR®
1030
62379-218EA 140.68 USD InStock
62379-218
VWR® Extra-Large LCD Digital Stopwatches
Timers Digital Stopwatches
Stopwatches are designed for routine quality control and experimental needs of labs, and for the exacting demands of timing industrial tests, enzyme analysis, or research results.

  • Traceable®
  • Quartz-crystal design

Large 1 cm (¹³/₃₂") LCD readout is visible indoors or outdoors.

Memory permits display of times after the event is over. Powerful battery provides four years of continuous use.

Readout is 9 hours, 59 minutes, 59 seconds, 99 hundredths. All stopwatches feature interval split, cumulative split, single action timing, time-out/time-in, continuous roll-over timing, and countdown alarm timing. Model 62379-207 has dual display that shows cumulative and interval splits and eight recallable memories; countdown timing function can be set to repeat on automatic cycles. Models 62379-229 and 62379-230 have decimal timing (99999.99 seconds, 999.999 minutes, 9.99999 hours), triple display, 500 recallable memories (including fastest, slowest, and average lap time), date display, pacer, and stroke measurement functions.

Stopwatches can be carried in hand or coat pocket, or worn around neck. Watertight construction gives positive protection against lab spills, even accidental submersion. Quartz-crystal design provides 100 times greater accuracy than mechanical watches.

Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Ordering information: Supplied with lanyard and two-year, replaceable 1.5 V silver-oxide battery.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty