Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Three-Button Stopwatch
Supplier: Avantor
VWR®
90027-82
62379-234EA 34.77 USD InStock
62379-234
VWR® Three-Button Stopwatch
Timers Digital Stopwatches
For easy-duty, occasional use where high precision is not a requirement.
  • Traceable®
  • Designed for occasional, easy-duty use
  • Multiple modes of functionality
  • Audible confirmation of button pressing
Unit times to 24 hours, with a resolution of 0.01 second for thirty minutes and one second thereafter. Modes include start/stop, time-out, cumulative split, and continuous roll-over timing. Also displays time and date. Provides a sound confirmation each time a button is pressed.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with lanyard and 1.5 V silver-oxide battery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty