Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Supplier: Avantor
VWR®
90010-36 90010-44
62379-460EA 44.27 USD InStock
62379-460 61161-350
VWR® Water-Resistant/Shock-Resistant and Waterproof/Shockproof Stopwatches
Timers Digital Stopwatches
Perfect for lab and plant use, this LCD digital stopwatch keeps time up to 24 hours.
  • Fine time-keeping resolution
  • Water-resistant, shock-resistant ABS plastic housing
  • Different models are color-coded for water resistance
  • Traceable®
Timing resolution is 0.01 seconds for the first 30 minutes.
Housed in a water-resistant/shock-resistant or waterproof/shockproof ABS plastic case. Water-resistant model is black. Waterproof model is yellow in color.
Continues timing to 23 hours, 59 minutes, 59 seconds with a 1 second resolution. Accuracy is 0.01%. Modes include single action, time-out, cumulative split, and continuous timing. Displays time-of-day in hours, minutes, and seconds, and features time-of-day alarm. Displays date and day of the week. Tactile-feel with 'click' switches provide positive action.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with lanyard, instructions, and two-year, 1.5 V silver-oxide battery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty