Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® All-Function Stopwatch with Countdown
Supplier: VWR
VWR®
90027-72
62379-517EA 122.7 USD InStock
62379-517
VWR® All-Function Stopwatch with Countdown
Timers Digital Stopwatches
All-purpose stopwatch is ideal for lab and plant use.
  • Traceable®
  • Recalls up to 300 memories
  • Stopwatch and chronograph capability
Stopwatch features include single action, time-in/time-out, continuous timing, cumulative split, interval split, 300 memories, and repetitive programmable countdown alarm times
Stopwatch times to 9 hours, 59 minutes, 59 seconds, 99 hundredths. When counting up, the three-line display simultaneously shows running time, cumulative split, and interval split. Interval split shows longest, shortest, and average time. Unit will recall 300 memories and display sample numbers of 1 to 300. Recalls memories both during and after event timing.
When zero is reached during countdown, the timer alarms and begins countdown again from programmed time. Repeat cycle continues automatically until stopped. Display always shows running time remaining to zero and cycles counted from 1 to 300.
ABS case with non-slip rubber sides is O-ring sealed, water- and shock-resistant.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Unit is supplied with a two-year 3 V lithium 'always on' battery, lanyard, detachable clip, and Velcro® fastener for vertical mounting.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty