Would you like to log in again?
VWR® Calendar/Thermometer Wall Clock
Supplier: Avantor
VWR®
90010-61
62379-519EA 147.53 USD InStock
62379-519
VWR® Calendar/Thermometer Wall Clock
Timers Wall Clocks
More than just a timekeeper, this wall clock integrates complete clock functions, a room temperature monitor, and a perpetual calendar.
  • Traceable®
  • Large Digits
  • Clock, calendar, and temperature functions
Time can be shown in a.m./p.m. format or 24-hour international time. Temperature Displays °C or °F. Calendar shows month, date, day of the week, and automatically adjusts for leap year. Large 5.4 cm (2¹/₈") LCD digits provide excellent visibility from up to 10.7 m (35') away.
Clock has two-tone color face and slim silhouette. Quartz crystal timing accuracy of 0.01%.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with wall mount and two AA alkaline batteries.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty