Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Timer Plus
Supplier: VWR
VWR®
93678-68
62379-604EA 70.28 USD InStock
62379-604
VWR® Timer Plus
Timers Digital Timers
Key chain timer is three timers in one — countdown alarm timer, stopwatch, and time-of-day clock.
  • Traceable®
  • Six-digit LCD display
  • Programmable timer functions
  • VWR six-year limited warranty
Ideal for catching test results and signaling time intervals and appointments.
Six-digit LCD always shows time in hours, minutes, and seconds. Countdown mode can be programmed in one-minute increments from 100 hours to 1 minute. When display reaches zero, a loud, beeping alarm sounds for 60 seconds. The alarm can be silenced manually. In stopwatch mode, it times up from 1 second to 99 hours and then repeats. The timer has a time-out feature in both countup and stopwatch modes. Time-of-day clock displays to the second with a.m./p.m. indicators and may be set in 24-hour military time.
Clip it to a button-hole, belt loop, or notebook.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with an eighteen-month continuous-use replaceable silver-oxide battery, keychain, lanyard, and magnet for placing on metal.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty