Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Stopwatch, ¹/₁₀ sec.
Supplier: Avantor
VWR®
141.0434-WO
62387-205EA 839.47 USD InStock
62387-205
VWR® Stopwatch, ¹/₁₀ sec.
Timers Analog Stopwatches
Compact, rugged, and simple to use, with time-out and safety reset features.
  • Timing to 0.1 second, plus time-out
  • Color-coded, push-button control
  • Shock-protected, pin-lever movement
  • ABS plastic case
All push-buttons are logically color-coded to minimize timing error and ensure efficient use — green for start, red for stop, and black for return to zero position.
Instant flyback function resets and starts immediately.
The 30-second dial is graduated in tenths of a second, and a 0 to 15 minute register is color-contrasted for easy reading.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty