Would you like to log in again?
Magnetic Spin Vanes for NextGen™ V Vial® Screw-Thread Sample Vials, Wheaton
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
903063 903061
62402-558CS 116.57 USD InStock
62402-558 62402-556
Magnetic Spin Vanes for NextGen™ V Vial® Screw-Thread Sample Vials, Wheaton
Stirrers Magnetic Stirring Bars
These fluoropolymer resin spin vanes have cross-mounted molded-in magnetic bars for V-vials.
These exceptionally sturdy vials have a conical interior bottom that provides total downward drainage for maximum retrieval of contents by syringe or micropipette.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty