Would you like to log in again?
REMOVAWELL 2 FBT1X12 CS320
Supplier: Thermo Fisher Scientific
Microlite* 2 Microtiter* Removawell* Strip^ 12-wells/strip, with flat-bottom well shape. Irradiated polystyrene. For luminescence applications. Microlite 2 is for applications requiring higher binding with the same high reflectivity and minimal crosstalk.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REMOVAWELL 2 FBT1X12 CS320 62403-102 7410 Case of 320 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty