Would you like to log in again?
Tissue Culture Rotators, Glas-Col®
Supplier: Glas-Col
099A RD4512
62404-006EA 2054.9 USD InStock
62404-006
Tissue Culture Rotators, Glas-Col®
Shakers and Mixers Rotating/Rocking Shakers
Ideal for continuous mixing of tissue culture specimens
Unique clamping device maintains motor position at any point within 110°. The variable speed control module with 1.2 m (4') safety cord allows for convenient operation at a distance or within controlled environments. Friction-drive test tube rotator head accommodates thirty-one 10 to 18 mm dia. tubes or sixteen 20 to 30 mm dia. tubes. Spring clips attach easily to rotator head to hold test tubes. Change heads with a simple thumbscrew operation. Containers remain visible during operation. Enameled steel body with a 1.3 cm (¹/₂") steel base plate with bumper feet.
Ordering information: Complete with test tube rotator head, 31 clips for ten 18 mm diameter tubes, and 16 clips for twenty 30 mm diameter tubes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty