Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Holder for Rugged Rotator, 10 to 30 mm Test Tubes, Glas-Col

Supplier: Glas-Col

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

099ARD65
62404-012EA 377.01 USD InStock
62404-012
Holder for Rugged Rotator, 10 to 30 mm Test Tubes, Glas-Col
Support Stands
Used with the Rugged Rotator for blood agitation, isotopic competitive protein binding assays and similar procedures.

  • Tubes can be inserted radially
  • Equipped with adjustable spring clips
  • Clips designed for side rather than top insertion of tubes

Furnished with 31 spring clips for 10 to 18 mm diameter tubes and 16 spring clips for 20 to 30 mm diameter tubes.

Ordering information: Holder with two sizes of spring clips.

Delivery information: 7 to 10 days after order.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty