Would you like to log in again?
Holder for Rugged Rotator, Test Tube, Glas-Col
Supplier: Glas-Col
099ARD10
62404-014EA 1147.16 USD InStock
62404-014
Holder for Rugged Rotator, Test Tube, Glas-Col
Support Stands
Frequently used with the Rugged Rotator for embedding tissue for electron microscopy.
  • Accepts a variety of glassware
  • Tubes easily inserted or removed
  • No slip disk design
The spaced disk with springs conveniently holds a combination of test tubes.
Ordering information: Holder only.
Delivery information: 7 to 10 days after order.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty