Would you like to log in again?
Glass Roller Culture Vessels, Large Neck, with Phenolic Caps, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences
348528 348522
62404-416CS 448.58 USD InStock
62404-416 62404-422
Glass Roller Culture Vessels, Large Neck, with Phenolic Caps, Wheaton®, DWK Life Sciences
Bottles Roller Bottles
Bottles feature a larger opening to provide easier access to the inside of the vessel.
The 51-400 caps have a shallow skirt and white styrene-butadiene rubber liner. Wheaton 33® low extractable borosilicate glass meets ASTM Type I Class A and USP Type I standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty