Would you like to log in again?
Corning® BioCoat® Fibrillar Collagen Cell Culture Inserts, Corning
Supplier: Corning
Corning® BioCoat®
354474 354474
62405-050CS 471.49 USD InStock
CACB354474 62405-050
Corning® BioCoat® Fibrillar Collagen Cell Culture Inserts, Corning
Microplates Insert Plates and Inserts
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
  • Storage: 2 to 8 °C
  • Shelf life: >= 3 months from date of shipment
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty