Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Corning® BioCoat™ Matrigel® Matrix Culture Dishes, Corning
Supplier: Corning
Corning® BioCoat®
354600 354600 354601 354601
62405-142CS 290.22 USD InStock
62405-142 62405-140 CACB354600 CACB354601
Corning® BioCoat™ Matrigel® Matrix Culture Dishes, Corning
Culture Dishes Cell Culture Dishes
Corning® BioCoat™ Matrigel® Matrix Cellware consists of Falcon® Multiwell Tissue Culture inserts, plates, and dishes treated with a uniform application of Corning® Matrigel® Basement Membrane Matrix to promote differentiation of a variety of cell types.
Corning® Matrigel® source: Engelbreth-Holm-Swarm (EHS) mouse tumor. Tested for ability to promote neurite outgrowth from chick dorsal root ganglia (in the absence of NGF). Tested and found negative for bacteria and fungi.
Cellware is stable for >=3 months from date of shipment when stored at –20°C (–4°F); keep frozen until use. Thin-layer cellware is stable for >=3 months when stored at 2 to 8°C (35.6 to 46.4°F). Inserts stable for >=3 months at –20°C (–4°F); keep frozen until use.
Convenient, ready-to-use cellware offers lot-to-lot consistency and reliable cell culture performance. Use Corning® BioCoat™ Matrigel® Matrix Cellware for the culture of muscle cells, epithelial cells, endothelial cells, and more.
Ordering information: Inserts are packaged six per 6-well plate and twelve per 24-well plate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty