Would you like to log in again?
Vented Screw Caps for Falcon® Flasks, Sterile, Corning
Supplier: Corning
Falcon®
354637 354638 354637 354639 354639 354638
62405-156CS 87.34 USD InStock
62405-156 62405-154 CACB354637 62405-152 CACB354639 CACB354638
Vented Screw Caps for Falcon® Flasks, Sterile, Corning
Closures Bottle Closures
PE with integral hydophobic 0.2 µm microporous membrane filter vent.
Vented caps minimize contamination associated with standard open incubation. Gases required for cell growth and metabolism pass freely through the vent, while microorganisms do not.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty