Would you like to log in again?
Corning® BioCoat® Collagen IV-Coated Flasks, Corning
Supplier: Corning
Corning® BioCoat® Corning®
354523 354528 354523 354528
62405-652CS 474.55 USD InStock
62405-652 62405-650 CA-CB354528 CA-CB354523
Corning® BioCoat® Collagen IV-Coated Flasks, Corning
Flasks Cell Culture Flasks
Collagen IV is a ubiquitous component of basement membranes. It plays a role in the regulation of cell growth, differentiation and adhesion, as well as tissue formation. Corning® BioCoat™ Collagen IV flasks are tissue culture vessels with a uniform application of mouse collagen type IV. These flasks are manufactured in a highly controlled environment and are rigorously tested to assure product consistency and performance.
Applications include promotion of cell attachment and spreading, cell differentiation and neurite outgrowth, increased proliferation of PC 12 cells, studies of effects of collagen IV on cell behaviour, cell adhesion assays, and serum-free or reduced serum culture.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty