Would you like to log in again?
Falcon® Cell Culture Inserts, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Falcon®
353182 353182 353092 353494 353495 353494 353492 353093 353180 353096 353103 353102 353091 353493 353181 353181 353096 353493 353095 353292 353095 353292 353495 353090 353492 353093 353097 353102 353092 353097 353090 353103 353091 353104 353180 353104
62406-169CS 326.28 USD InStock
CA62406-178 62406-169 CA62406-177 CA62406-179 CA62406-196 CA62406-174 62406-164 CA62406-173 CA62406-176 CA62406-198 CA62406-175 CA62406-181 62406-171 CA62406-180 62406-172 62406-173 62406-196 62406-174 62406-170 62406-179 CA62406-169 62406-175 CA62406-163 62406-176 62406-198 62406-177 62406-178 CA62406-164 CA62404-550 62404-550 CA62406-170 CA62406-172 CA62406-171 62406-163 62406-180 62406-181
Falcon® Cell Culture Inserts, Corning
Microplates Insert Plates and Inserts
Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane.
  • Low protein binding PET membrane
  • Sterilized by gamma irradiation
  • A wide variety of configurations including 6, 12 and 24 well
  • A broad selection of membrane pore sizes, 0.4, 1.0, 3.0 and 8.0 µm diameter
  • Packed in individual blister packs, 48 inserts/case
  • Non-Tissue Culture-treated insert housings prevent promiscuous growth of cells on the insert walls
  • Innovative hanging design facilitates pipetting and allows for co-culture
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty