Would you like to log in again?
Falcon® Cell Culture Insert Companion Plates, Corning
Supplier: Corning
Falcon®
355467 353503 353504 353503 353504 353502 355467 353502
62406-187CS 224.49 USD InStock
CA62406-189 CA62406-185 62406-187 CA62406-187 62406-189 62405-150 62406-185 CA-CB355467
Falcon® Cell Culture Insert Companion Plates, Corning
Microplates Insert Plates and Inserts
PS, tissue culture-treated, sterile, non-pyrogenic, with lid. Specifically designed for use with Falcon® or Corning® BioCoat™ cell culture inserts, so that evaporation and contamination due to improper lid fit is eliminated.
  • Reagents can be added quickly and consistently for timed experiments
  • Aspiration from the well is easier, reducing the risk of contamination
  • Media cannot wick up between the insert and the well wall
  • Low-evaporation lid reduces evaporation and contamination
In the 'Feeding Position' pipette access is improved for fluid handling on the basolateral side. In the 'Incubation Position' cell culture inserts remain locked in position in their companion plate wells.
Note: May be used with or without cell culture inserts.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty