Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Corning® BioCoat® Poly-D-Lysine Culture Dishes, Corning
Supplier: Corning
Corning® BioCoat®
356468 356468 356469 354467 356467 354469 354468 354550 354550 354467 354469 356469 356467 354468
62406-048CS 110.52 USD InStock
62406-048 62406-038 62406-044 CA-CB354467 CA-CB354468 CA-CB354469 62406-702 CA-CB354550 CACB356467A CACB356468A CACB356469A 62406-050 62406-040 62406-042
Corning® BioCoat® Poly-D-Lysine Culture Dishes, Corning
Culture Dishes Cell Culture Dishes
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
  • Attachment and spreading of a variety of cell types
  • Cell differentiation and neurite outgrowth
  • Attachment of fastidious transfected cell lines
  • Support survival of primary neurons in culture
  • Serum-free or reduced-serum culture
  • Storage: 2 to 8 °C under dry conditions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty